Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285785.1 | NP_001272714.1 | 6 | 937 | ||
NM_146011.2 | NP_666123.2 | 5 | 376 | |||
GenBank mRNA | AK049245.1 | - | 2 | 157 | ||
AK080955.1 | - | 4 | 1071 | |||
AK083474.1 | - | 2 | 286 | |||
AK089051.1 | - | 5 | 328 | |||
AK167738.1 | - | 6 | 514 | |||
BC024535.1 | - | 5 | 371 | 4987970 | ||
BC027374.1 | - | 5 | 347 | 4947306 | ||
DQ498128.1 | - | 6 | 785 |