Search Thermo Fisher Scientific
Gene Symbol: Mgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145137.2 | NP_660119.1 | 10 | 1105 | ||
GenBank mRNA | AK085751.1 | - | 8 | 1109 | ||
AY103461.1 | - | 10 | 1042 | |||
BC106833.1 | - | 9 | 1031 | 40044847 | ||
BC110475.2 | - | 9 | 1031 | 40045765 | ||
BC111520.1 | - | 9 | 1031 | 40042306 | ||
EU010011.1 | - | 9 | 1184 |