Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015099.2 | NP_001015099.2 | 15 | 2100 | ||
NM_001167963.1 | NP_001161435.1 | 16 | 2169 | |||
NM_001167964.1 | NP_001161436.1 | 14 | 1818 | |||
GenBank mRNA | AK031333.1 | - | 14 | 1678 | ||
AK033513.1 | - | 16 | 2113 | |||
AK129332.1 | - | 15 | 2003 | |||
AK138847.1 | - | 2 | 1099 | |||
BC086455.1 | - | 15 | 1973 | 30458160 |