Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029786.1 | - | 4 | 2060 | ||
AK132195.1 | - | 4 | 3468 | |||
AK162464.1 | - | 3 | 2135 | |||
BC003865.1 | - | 3 | 262 | |||
BC072599.1 | - | 4 | 3479 | |||
BC079861.1 | - | 4 | 3466 | 6834767 |