Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289550.1 | NP_001276479.1 | 9 | 1113 | ||
NM_001289551.1 | NP_001276480.1 | 9 | 1109 | |||
NM_001289552.1 | NP_001276481.1 | 8 | 970 | |||
NM_001289553.1 | NP_001276482.1 | 8 | 997 | |||
GenBank mRNA | AK003170.1 | - | 9 | 947 | ||
AK041233.1 | - | 9 | 942 | |||
AK085042.1 | - | 8 | 839 | |||
AK087114.1 | - | 9 | 944 | |||
AK156423.1 | - | 9 | 948 | |||
AK158409.1 | - | 8 | 822 | |||
BC056362.1 | - | 8 | 2222 | 6816930 | ||
DQ143891.1 | - | 8 | 998 | |||
DQ143892.1 | - | 7 | 784 | |||
DQ143893.1 | - | 7 | 791 | |||
DQ143894.1 | - | 7 | 786 | |||
DQ143895.1 | - | 7 | 1202 | |||
DQ143896.1 | - | 7 | 1163 | |||
DQ143897.1 | - | 7 | 1241 | |||
DQ143898.1 | - | 7 | 1190 | |||
DQ143899.1 | - | 6 | 1317 | |||
DQ143900.1 | - | 8 | 1010 | |||
L22482.1 | - | 8 | 1074 |