Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153789.3 | NP_722484.2 | 7 | 1535 | ||
GenBank mRNA | AK034887.1 | - | 7 | 1535 | ||
AK040836.1 | - | 7 | 1402 | |||
AK159478.1 | - | 7 | 1536 | |||
AK171888.1 | - | 7 | 1536 | |||
AY434449.1 | - | 7 | 1320 | |||
BC010206.1 | - | 7 | 1391 | 3964381 |