Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009378.3 | NP_033404.1 | 1 | 564 | ||
GenBank mRNA | AK044928.1 | - | 1 | 496 | ||
AK089479.1 | - | 1 | 565 | |||
BC019154.1 | - | 1 | 489 | 5065481 | ||
CT010375.1 | - | 1 | 322 | |||
X14432.1 | - | 1 | 480 |