Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113417.1 | NP_001106888.1 | 8 | 1032 | ||
NM_009380.3 | NP_033406.1 | 5 | 1118 | |||
GenBank mRNA | AB221613.1 | - | 5 | 885 | ||
AK133662.1 | - | 5 | 615 | |||
AK158826.1 | - | 8 | 1033 | |||
BC089035.1 | - | 5 | 1118 | 30695885 | ||
BC119552.1 | - | 5 | 1055 | 40057420 | ||
BC119553.1 | - | 5 | 1059 | 40057421 | ||
S62756.1 | - | 7 | 902 |