Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198605.3 | NP_941007.1 | 6, 7 | 982 | ||
GenBank mRNA | AK089223.1 | - | 6, 7 | 983 | ||
AK166171.1 | - | 6, 7 | 973 | |||
BC117500.1 | - | 6, 7 | 974 | 40105921 | ||
BC117501.1 | - | 6, 7 | 974 | 40105920 |