Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113404.1 | NP_001106875.1 | 1 | 110 | ||
NM_011648.5 | NP_035778.3 | 1 | 110 | |||
GenBank mRNA | AK014519.1 | - | 1 | 109 | ||
AK021020.1 | - | 1 | 104 | |||
AK029084.1 | - | 1 | 76 | |||
AK041086.1 | - | 1 | 87 | |||
BB645263.1 | - | 1 | 110 | |||
BC028754.1 | - | 1 | 65 | |||
BC086691.1 | - | 1 | 39 | 30682686 | ||
BC092523.1 | - | 1 | 52 | 30650002 | ||
U02601.1 | - | 1 | 92 | |||
U02602.1 | - | 1 | 92 |