Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011669.3 | NP_035799.1 | 4, 5 | 856 | ||
GenBank mRNA | AF022792.1 | - | 1, 2 | 125 | ||
AF441835.1 | - | 3, 4 | 517 | |||
AK006739.1 | - | 4, 5 | 906 | |||
AK035629.1 | - | 4, 5 | 697 | |||
AK167153.1 | - | 4, 5 | 696 | |||
BC049274.1 | - | 2, 3 | 523 | 4500832 | ||
BC068136.1 | - | 4, 5 | 856 | 30363916 |