Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009513.2 | NP_033539.1 | 3 | 319 | ||
GenBank mRNA | AK138191.1 | - | 3 | 250 | ||
AK153741.1 | - | 2 | 227 | |||
AK160941.1 | - | 3 | 305 | |||
AU080650.1 | - | 3 | 405 | |||
BC052019.1 | - | 2 | 142 | 5708003 | ||
D83206.1 | - | 3 | 272 |