Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183017.2 | NP_898838.2 | 12 | 1661 | ||
GenBank mRNA | AK084879.1 | - | 12 | 1628 | ||
AK134345.1 | - | 12 | 1628 | |||
AK143496.1 | - | 11 | 1494 | |||
AK150116.1 | - | 12 | 1627 | |||
AM690756.1 | - | 12 | 1614 | |||
BC055368.1 | - | 12 | 1626 | 30055801 |