Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Onecut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194268.2 | NP_919244.2 | 1, 2 | 1241 | ||
GenBank mRNA | AY242995.1 | - | 1, 2 | 1241 | ||
BC100750.1 | - | 2, 3 | 738 | |||
BC100755.1 | - | 2, 3 | 738 | |||
BC103668.1 | - | 2, 3 | 1074 | 40046651 |