Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081259.1 | NP_001074728.1 | 10 | 1760 | ||
GenBank mRNA | AK036315.1 | - | 9 | 1357 | ||
AK044206.1 | - | 2 | 140 | |||
AK165872.1 | - | 10 | 1760 | |||
BC024751.1 | - | 5 | 738 | |||
BC157992.1 | - | 10 | 1728 | 9007305 |