Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160139.1 | NP_001153611.1 | 15 | 3015 | ||
NM_023872.3 | NP_076361.1 | 14 | 2958 | |||
GenBank mRNA | AF263836.1 | - | 14 | 2433 | ||
AK020325.1 | - | 1 | 90 | |||
AK033079.1 | - | 8 | 1516 | |||
AK147264.1 | - | 15 | 3018 | |||
AY679158.1 | - | 14 | 2597 |