Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009555.2 | NP_033581.2 | 6 | 1113 | ||
GenBank mRNA | AK049894.1 | - | 6 | 1117 | ||
BC061205.1 | - | 6 | 1082 | 30248495 | ||
BC065097.1 | - | 6 | 1106 | |||
D10632.1 | - | 2 | 795 |