Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172659.2 | NP_766247.2 | 8 | 1239 | ||
GenBank mRNA | AK079650.1 | - | 8 | 1183 | ||
AK089246.1 | - | 8 | 1231 | |||
AK137441.1 | - | 7 | 1454 | |||
AK150104.1 | - | 8 | 1192 | |||
AK163445.1 | - | 8 | 1197 | |||
AK166312.1 | - | 8 | 1197 | |||
AK172411.1 | - | 7 | 1463 | |||
BC025498.1 | - | 5 | 1823 | |||
BC058210.1 | - | 7 | 1448 | |||
BC145480.1 | - | 8 | 1165 |