Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011772.2 | NP_035902.2 | 7 | 1108 | ||
GenBank mRNA | AB017615.1 | - | 6 | 1515 | ||
AK089522.1 | - | 7 | 942 | |||
AK170453.1 | - | 8 | 1032 | |||
BC141286.1 | - | 6 | 1206 | 9055898 | ||
BC145614.1 | - | 6 | 1192 |