Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144892.1 | NP_659141.1 | 8 | 1709 | ||
GenBank mRNA | AK049242.1 | - | 5 | 1113 | ||
AK173226.1 | - | 6 | 1551 | |||
BC017152.1 | - | 8 | 1709 | 4511696 | ||
BC022624.1 | - | 1 | 187 |