Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172676.2 | NP_766264.2 | 4, 5 | 708 | ||
GenBank mRNA | AK053048.1 | - | 4, 5 | 662 | ||
AK053884.1 | - | 2, 3 | 289 | |||
AK164204.1 | - | 4, 5 | 748 | |||
AK171151.1 | - | 4, 5 | 714 | |||
BC019475.1 | - | 4, 5 | 718 | |||
BC052921.1 | - | 4, 5 | 708 | 6311715 |