Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172678.3 | NP_766266.3 | 7 | 893 | ||
GenBank mRNA | AK028571.1 | - | 6 | 1607 | ||
AK031820.1 | - | 7 | 825 | |||
AK075984.1 | - | 7 | 812 | |||
AK145423.1 | - | 7 | 801 | |||
AK178887.1 | - | 1 | 17 | |||
AK179129.1 | - | 1 | 17 | |||
AK203069.1 | - | 1 | 17 | |||
AK206153.1 | - | 1 | 107 | |||
AK208945.1 | - | 1 | 17 | |||
AK215567.1 | - | 2 | 183 | |||
AK217282.1 | - | 1 | 117 |