Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008725.2 | NP_032751.1 | 1 | 150 | ||
GenBank mRNA | AK147180.1 | - | 1 | 154 | ||
BC089615.1 | - | 1 | 150 | 6755676 | ||
KC526925.1 | - | 1 | 105 | |||
KC526926.1 | - | 1 | 105 | |||
KC526927.1 | - | 1 | 105 |