Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008725.2 | NP_032751.1 | 1, 2 | 209 | ||
GenBank mRNA | AK147180.1 | - | 1, 2 | 213 | ||
BC089615.1 | - | 1, 2 | 209 | 6755676 |