Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172704.3 | NP_766292.2 | 12, 13 | 1356 | ||
GenBank mRNA | AK053156.1 | - | 12, 13 | 1338 | ||
AK077544.1 | - | 3, 4 | 290 | |||
AK171781.1 | - | 12, 13 | 1337 | |||
AK172542.1 | - | 12, 13 | 1337 | |||
BC079852.1 | - | 12, 13 | 1326 | |||
BC085257.1 | - | 12, 13 | 1330 | 5414963 |