Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013378.2 | NP_001013396.2 | 5, 6 | 1258 | ||
NM_001115149.1 | NP_001108621.1 | 5, 6 | 1258 | |||
NM_001115150.1 | NP_001108622.1 | 4, 5 | 1089 | |||
NM_001115151.1 | NP_001108623.1 | 4, 5 | 1089 | |||
NM_001115153.1 | NP_001108625.1 | 6, 7 | 1435 | |||
NM_001286824.1 | NP_001273753.1 | 5, 6 | 1216 | |||
NM_001286825.1 | NP_001273754.1 | 5, 6 | 1200 | |||
GenBank mRNA | AK050969.1 | - | 5, 6 | 1242 | ||
AK088787.1 | - | 4, 5 | 955 | |||
AK145409.1 | - | 6, 7 | 1333 | |||
AK146820.1 | - | 5, 6 | 1083 | |||
AK161654.1 | - | 4, 5 | 936 | |||
AK168702.1 | - | 5, 6 | 1083 | |||
AK168981.1 | - | 5, 6 | 1110 | |||
BC028849.1 | - | 5, 6 | 1152 | |||
BC032291.1 | - | 2, 3 | 3228 | |||
BC086664.1 | - | 5, 6 | 1100 |