Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177713.1 | NP_001171184.1 | 4 | 868 | ||
NM_175475.3 | NP_780684.1 | 4 | 919 | |||
GenBank mRNA | AK028272.1 | - | 2 | 403 | ||
AK137124.1 | - | 3 | 1194 | |||
AK166211.1 | - | 3 | 457 | |||
AY134662.1 | - | 4 | 855 | |||
BC059246.1 | - | 4 | 868 | 6400154 |