Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081239.2 | NP_001074708.2 | 4 | 640 | ||
GenBank mRNA | AK086983.1 | - | 1 | 53 | ||
AY998482.1 | - | 4 | 640 |