Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172742.2 | NP_766330.2 | 10 | 1127 | ||
GenBank mRNA | BC041672.1 | - | 10 | 1099 | ||
BC055074.1 | - | 8 | 791 | 30052208 | ||
BC058196.1 | - | 3 | 229 | |||
CF734341.1 | - | 5 | 506 |