Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026964.3 | NP_081240.1 | 2 | 169 | ||
GenBank mRNA | AK007589.1 | - | 2 | 170 | ||
AK088037.1 | - | 2 | 180 | |||
BC049678.1 | - | 2 | 169 | 6595832 | ||
BC050938.1 | - | 1 | 147 | 6819562 |