Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181594.3 | NP_853625.1 | 18, 19 | 2738 | ||
GenBank mRNA | BC053081.1 | - | 18, 19 | 2668 | 5704584 |