Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172775.2 | NP_766363.2 | 29 | 5624 | ||
GenBank mRNA | AB072381.1 | - | 29 | 5468 | ||
AK129133.1 | - | 18 | 2621 | |||
BC026974.1 | - | 5 | 649 | |||
BC150701.1 | - | 28 | 5416 | 9087612 |