Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198111.2 | NP_932779.2 | 13 | 5628 | ||
GenBank mRNA | AK129115.1 | - | 1 | 1087 | ||
AK147248.1 | - | 13 | 5629 | |||
AK147371.1 | - | 13 | 5629 | |||
AK162929.1 | - | 2 | 2023 | |||
DQ233645.1 | - | 13 | 5684 | |||
DQ233646.1 | - | 12 | 5478 |