Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024285.2 | NP_077247.1 | 8 | 1187 | ||
GenBank mRNA | AF124510.1 | - | 7 | 1045 | ||
AF204174.1 | - | 8 | 1185 | |||
AK046765.1 | - | 3 | 282 | |||
BC132018.1 | - | 7 | 1077 | 40130295 | ||
BC132044.1 | - | 7 | 1077 | 40130321 |