Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081128.3 | NP_001074597.1 | 28 | 3196 | ||
GenBank mRNA | AK085987.1 | - | 3 | 495 | ||
BC145683.1 | - | 28 | 3083 | 40130986 | ||
BC145685.1 | - | 28 | 3083 | 40130988 |