Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 7 | 667 | ||
NM_001253784.1 | NP_001240713.1 | 6 | 645 | |||
NM_001281803.1 | NP_001268732.1 | 5 | 649 | |||
NM_054046.5 | NP_473387.1 | 5 | 558 | |||
GenBank mRNA | AK028726.1 | - | 6 | 617 | ||
AK034028.1 | - | 524 | ||||
AK046271.1 | - | 5 | 518 | |||
AK050405.1 | - | 6 | 616 | |||
AK081295.1 | - | 5 | 548 | |||
AK086174.1 | - | 5 | 519 | |||
AK087778.1 | - | 5 | 529 | |||
AK129019.1 | - | 5 | 600 | |||
AK133979.1 | - | 5 | 532 | |||
AK150334.1 | - | 5 | 494 | |||
AK150994.1 | - | 5 | 494 | |||
AK164147.1 | - | 5 | 530 | |||
BC003306.1 | - | 7 | 618 | 3154712 | ||
BY713285.1 | - | 5 | 559 | |||
CD545768.1 | - | 1 | 44 | |||
CX562217.1 | - | 5 | 611 |