Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183175.4 | NP_898998.2 | 2 | 106 | ||
GenBank mRNA | BC115632.1 | - | 2 | 91 | 40102920 | |
BC115633.1 | - | 2 | 91 | 40102924 | ||
DQ002401.1 | - | 2 | 106 |