Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011836.3 | NP_035966.2 | 21 | 3657 | ||
GenBank mRNA | AF079520.1 | - | 21 | 3680 | ||
AF083372.1 | - | 21 | 3594 | |||
AK132948.1 | - | 18 | 2644 | |||
BC005535.1 | - | 4 | 402 | |||
BC096366.1 | - | 21 | 3657 | 2865309 |