Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081970.1 | NP_001075439.1 | 4 | 667 | ||
GenBank mRNA | AK036408.1 | - | 4 | 687 | ||
AK046043.1 | - | 4 | 796 | |||
BC129972.1 | - | 4 | 667 | 40090463 |