Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291275.1 | NP_001278204.1 | 1 | 260 | ||
NM_001291276.1 | NP_001278205.1 | 1 | 260 | |||
NM_011870.5 | NP_036000.1 | 1 | 260 | |||
GenBank mRNA | AB006463.1 | - | 1 | 41 | ||
AF173010.1 | - | 1 | 39 | |||
AK010345.1 | - | 1 | 57 | |||
AK027915.1 | - | 1 | 72 | |||
AK088450.1 | - | 1 | 75 | |||
AK152805.1 | - | 1 | 92 | |||
AK165472.1 | - | 1 | 45 | |||
BB843350.1 | - | 1 | 39 | |||
BC003714.1 | - | 1 | 46 | 3599986 | ||
BC054385.1 | - | 1 | 65 | 6476137 | ||
CF104297.1 | - | 1 | 268 |