Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081656.2 | NP_001075125.1 | 5 | 1484 | ||
GenBank mRNA | DQ839448.1 | - | 5 | 1484 | ||
GQ414760.1 | - | 5 | 1484 |