Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dytn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081658.1 | NP_001075127.1 | 5 | 610 | ||
GenBank mRNA | AK158076.1 | - | 3 | 3349 | ||
DQ443727.1 | - | 5 | 610 |