Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163566.1 | NP_001157038.1 | 8 | 2160 | ||
GenBank mRNA | BC043114.1 | - | 7 | 1848 | ||
BC062128.1 | - | 1 | 18 |