Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163566.1 | NP_001157038.1 | 11, 12 | 3577 | ||
GenBank mRNA | AK031068.1 | - | 2, 3 | 296 | ||
BC043114.1 | - | 10, 11 | 3265 | |||
BC062128.1 | - | 4, 5 | 1435 |