Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177363.3 | NP_796337.1 | 4 | 730 | ||
GenBank mRNA | AK036904.1 | - | 4 | 731 | ||
AK089206.1 | - | 723 | ||||
AK155804.1 | - | 4 | 738 | |||
AK170277.1 | - | 3 | 661 | |||
DQ973493.1 | - | 4 | 682 |