Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 9 | 2064 | ||
GenBank mRNA | AF338472.1 | - | 8 | 1222 | ||
AF496547.1 | - | 9 | 1294 | |||
AK051262.1 | - | 9 | 2065 | |||
AK122276.1 | - | 9 | 1712 |