Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 11, 12 | 1126 | ||
GenBank mRNA | AK008141.1 | - | 11, 12 | 1129 | ||
AK011002.1 | - | 11, 12 | 1127 | |||
AK150067.1 | - | 11, 12 | 1127 | |||
AK159379.1 | - | 11, 12 | 1125 | |||
AK166255.1 | - | 11, 12 | 1128 | |||
AK166507.1 | - | 11, 12 | 1127 | |||
AK167209.1 | - | 11, 12 | 1127 | |||
AK168710.1 | - | 11, 12 | 1109 | |||
AK176968.1 | - | 2, 3 | 96 | |||
BC007467.1 | - | 10, 11 | 1065 | 3497714 | ||
D87115.1 | - | 11, 12 | 1139 | |||
X93150.1 | - | 9, 10 | 861 |