Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252387.1 | NP_001239316.1 | 2 | 1486 | ||
NM_182807.3 | NP_877959.2 | 3 | 1698 | |||
GenBank mRNA | AK032483.1 | - | 2 | 568 | ||
AK139403.1 | - | 2 | 1570 | |||
AK140763.1 | - | 2 | 1449 | |||
AK164201.1 | - | 2 | 578 | |||
AY325121.1 | - | 3 | 385 | |||
BC027082.1 | - | 3 | 1584 | 5360443 |