Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164089.1 | NP_001157561.1 | 2 | 312 | ||
NM_012003.2 | NP_036133.1 | 2 | 297 | |||
GenBank mRNA | AF071316.1 | - | 2 | 299 | ||
AF240179.1 | - | 1 | 117 | |||
AK008474.1 | - | 2 | 303 | |||
AK010417.1 | - | 2 | 307 | |||
AK010726.1 | - | 2 | 282 | |||
AK013155.1 | - | 2 | 265 | |||
AK019122.1 | - | 2 | 289 | |||
BC003724.1 | - | 1 | 178 | 3593979 | ||
BG085353.2 | - | 2 | 297 | |||
CJ165979.1 | - | 2 | 293 |