Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfasc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160316.1 | NP_001153788.1 | 2 | 256 | ||
NM_001160317.1 | NP_001153789.1 | 4 | 492 | |||
NM_001160318.1 | NP_001153790.1 | 4 | 593 | |||
NM_182716.4 | NP_874385.1 | 5 | 510 | |||
GenBank mRNA | AJ543322.1 | - | 5 | 479 | ||
AK052440.1 | - | 3 | 385 | |||
AK129207.1 | - | 4 | 593 | |||
AK138636.1 | - | 4 | 407 | |||
BB659942.1 | - | 3 | 385 |
Set Membership: |
G-protein coupled receptor |